Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpr16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043212.3 | NP_001036677.1 | 2 - 3 | 958 | 67 | |
NM_001300240.1 | NP_001287169.1 | 4 - 5 | 1800 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |