Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glob3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141340.3 | NP_649597.3 | 2 - 3 | 490 | 104 | |
GenBank mRNA | BT022666.1 | - | 2 - 3 | 490 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |