Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104224.2 | NP_001097694.1 | 1 - 2 | 176 | 67 | |
NM_141354.3 | NP_649611.2 | 1 - 2 | 176 | 67 | ||
GenBank mRNA | AY069336.1 | - | 1 - 2 | 144 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |