Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141361.2 | NP_649618.1 | 2 - 3 | 610 | 58 | |
GenBank mRNA | BT100152.4 | - | 2 - 3 | 514 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |