Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osi4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275376.1 | NP_001262305.1 | 1 - 2 | 589 | 91 | |
NM_141366.2 | NP_649623.2 | 1 - 2 | 589 | 91 | ||
GenBank mRNA | AY119041.1 | - | 1 - 2 | 589 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster |