Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osi18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141382.3 | NP_649639.1 | 1 - 2 | 547 | 64 | |
GenBank mRNA | AY070982.1 | - | 1 - 2 | 545 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |