Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141435.3 | NP_649692.2 | 2 - 3 | 501 | 68 | |
GenBank mRNA | BT128902.1 | - | 2 - 3 | 469 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |