Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir84a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141463.2 | NP_649720.2 | 3 - 4 | 823 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |