Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141477.2 | NP_649734.2 | 5 - 6 | 1461 | 100 | |
NM_169192.3 | NP_731174.1 | 4 - 5 | 1028 | 100 | ||
NM_169193.3 | NP_731175.2 | 4 - 5 | 1028 | 100 | ||
GenBank mRNA | AF233093.1 | - | 4 - 5 | 1019 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |