Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ScsbetaA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104243.3 | NP_001097713.1 | 3 - 4 | 390 | 97 | |
NM_001170087.2 | NP_001163558.1 | 3 - 4 | 390 | 97 | ||
NM_001170088.2 | NP_001163559.1 | 4 - 5 | 362 | 97 | ||
NM_001202268.3 | NP_001189197.2 | 4 - 5 | 362 | 97 | ||
NM_001202269.2 | NP_001189198.2 | 3 - 4 | 390 | 97 | ||
NM_001316546.1 | NP_001303475.1 | 3 - 4 | 390 | 97 | ||
NM_001316547.1 | NP_001303476.1 | 4 - 5 | 362 | 97 | ||
NM_141589.4 | NP_649846.2 | 4 - 5 | 362 | 97 | ||
GenBank mRNA | AY058331.1 | - | 3 - 4 | 362 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |