Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ScsbetaA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104243.3 | NP_001097713.1 | 7 - 8 | 1419 | 83 | |
NM_001170087.2 | NP_001163558.1 | 7 - 8 | 1422 | 83 | ||
NM_001170088.2 | NP_001163559.1 | 8 - 9 | 1394 | 83 | ||
NM_141589.4 | NP_649846.2 | 8 - 9 | 1391 | 83 | ||
GenBank mRNA | AY058331.1 | - | 7 - 8 | 1394 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |