Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HP1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141621.2 | NP_649878.1 | 1 | 402 | 138 | |
GenBank mRNA | BT100074.4 | - | 1 | 346 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |