Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nmdyn-D7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141669.3 | NP_649926.2 | 1 - 2 | 274 | 78 | |
GenBank mRNA | BT150178.1 | - | 1 - 2 | 272 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |