Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: topi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141688.1 | NP_649945.1 | 2 - 3 | 1501 | 71 | |
GenBank mRNA | BT150247.1 | - | 2 - 3 | 1498 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |