Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bruce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275531.2 | NP_001262460.1 | 20 - 21 | 8876 | 71 | |
NM_001275532.2 | NP_001262461.1 | 18 - 19 | 8504 | 71 | ||
NM_001275533.2 | NP_001262462.1 | 19 - 20 | 8573 | 71 | ||
NM_001275534.2 | NP_001262463.1 | 19 - 20 | 8576 | 71 | ||
NM_141738.5 | NP_649995.2 | 19 - 20 | 8576 | 71 | ||
GenBank mRNA | AF517634.1 | - | 19 - 20 | 8573 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |