Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap-H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141757.2 | NP_650014.2 | 4 - 5 | 1615 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |