Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap-H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141757.2 | NP_650014.2 | 7 - 8 | 2584 | 88 | |
NM_169358.2 | NP_731502.2 | 7 - 8 | 2545 | 88 | ||
NM_169359.2 | NP_731503.2 | 7 - 8 | 2581 | 88 | ||
NM_206471.2 | NP_996193.1 | 6 - 7 | 2310 | 88 | ||
GenBank mRNA | BT001334.1 | - | 6 - 7 | 2310 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |