Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cad86C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260102.1 | NP_001247031.1 | 5 - 6 | 1413 | 56 | |
NM_001260103.1 | NP_001247032.1 | 4 - 5 | 1329 | 56 | ||
NM_001260104.1 | NP_001247033.1 | 5 - 6 | 1413 | 56 | ||
NM_001260105.1 | NP_001247034.1 | 5 - 6 | 2069 | 56 | ||
NM_176458.4 | NP_788635.3 | 5 - 6 | 2069 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |