Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KP78a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141809.3 | NP_650066.1 | 2 | 2595 | 64 |
Gene Symbol: KP78b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170116.2 | NP_001163587.1 | 2 | 2595 | 64 | |
GenBank mRNA | BT031339.1 | - | 2 | 2595 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |