Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ninaG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260118.1 | NP_001247047.1 | 6 - 7 | 1660 | 97 | |
NM_141813.2 | NP_650070.1 | 5 - 6 | 1708 | 97 | ||
GenBank mRNA | AY118818.1 | - | 6 - 7 | 1660 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |