Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141863.3 | NP_650120.1 | 3 - 4 | 645 | 63 | |
NM_169434.2 | NP_731639.1 | 3 - 4 | 570 | 63 | ||
GenBank mRNA | AY089465.1 | - | 3 - 4 | 727 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |