Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141866.2 | NP_650123.1 | 2 - 3 | 568 | 95 | |
GenBank mRNA | BT016061.1 | - | 2 - 3 | 535 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |