Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tango9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316533.1 | NP_001303462.1 | 2 - 3 | 511 | 69 | |
NM_141879.3 | NP_650136.1 | 2 - 3 | 511 | 69 | ||
GenBank mRNA | AY058745.1 | - | 2 - 3 | 466 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |