Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ect3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141885.3 | NP_650142.1 | 4 - 5 | 1021 | 70 | |
GenBank mRNA | AY119565.1 | - | 4 - 5 | 992 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |