Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD-R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169461.3 | NP_731688.2 | 3 - 4 | 2131 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |