Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD-R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169461.3 | NP_731688.2 | 4 - 5 | 3461 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |