Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yrt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144572.2 | NP_001138044.1 | 4 - 5 | 2329 | 85 | |
NM_001144573.2 | NP_001138045.1 | 4 - 5 | 2329 | 85 | ||
NM_001144574.2 | NP_001138046.1 | 4 - 5 | 2329 | 85 | ||
NM_142034.2 | NP_650291.1 | 4 - 5 | 2329 | 85 | ||
GenBank mRNA | BT015265.1 | - | 4 - 5 | 2319 | 85 | |
EF078694.1 | - | 4 - 5 | 1617 | 85 | ||
EF078695.1 | - | 4 - 5 | 1617 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |