Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dip-B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300387.1 | NP_001287316.1 | 2 - 3 | 354 | 54 | |
NM_142061.3 | NP_650318.1 | 3 - 4 | 575 | 54 | ||
NM_169557.2 | NP_731863.1 | 2 - 3 | 479 | 54 | ||
NM_169558.2 | NP_731864.1 | 3 - 4 | 588 | 54 | ||
GenBank mRNA | AY051926.1 | - | 3 - 4 | 553 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |