Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MetRS-m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275635.2 | NP_001262564.1 | 5 - 6 | 1341 | 83 | |
NM_001316578.1 | NP_001303507.1 | 5 - 6 | 1341 | 83 | ||
NM_142091.5 | NP_650348.1 | 5 - 6 | 1341 | 83 | ||
GenBank mRNA | AY069128.1 | - | 5 - 6 | 1312 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |