Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCR-C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142144.3 | NP_650401.1 | 4 - 5 | 662 | 76 | |
NM_169607.2 | NP_731954.1 | 3 - 4 | 734 | 76 | ||
GenBank mRNA | AY058243.1 | - | 3 - 4 | 615 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |