Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170138.1 | NP_001163609.1 | 8 - 9 | 1274 | 153 | |
NM_001170140.1 | NP_001163611.1 | 7 - 8 | 977 | 153 | ||
NM_001170142.1 | NP_001163613.1 | 8 - 9 | 1208 | 153 | ||
NM_142166.2 | NP_650423.1 | 8 - 9 | 1364 | 153 | ||
GenBank mRNA | AY071198.1 | - | 8 - 9 | 1364 | 153 | |
BT081982.1 | - | 8 - 9 | 1266 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |