Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sxe2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300423.1 | NP_001287352.1 | 4 - 5 | 593 | 57 | |
NM_142265.3 | NP_650522.1 | 2 - 3 | 549 | 57 | ||
GenBank mRNA | AY058391.1 | - | 2 - 3 | 549 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |