Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mat89Ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142273.4 | NP_650530.2 | 2 - 3 | 1490 | 114 | |
GenBank mRNA | BT001511.1 | - | 2 - 3 | 1464 | 114 | |
BT016113.1 | - | 2 - 3 | 1462 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |