Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275724.1 | NP_001262653.1 | 1 - 2 | 276 | 102 | |
NM_142327.3 | NP_650584.1 | 1 - 2 | 276 | 102 | ||
GenBank mRNA | AY051443.1 | - | 1 - 2 | 254 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Drosophila melanogaster |