Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275749.1 | NP_001262678.1 | 4 - 5 | 2694 | 72 | |
NM_001275750.1 | NP_001262679.1 | 5 - 6 | 2619 | 72 | ||
NM_142388.3 | NP_650645.1 | 5 - 6 | 2709 | 72 | ||
NM_169768.2 | NP_732256.1 | 4 - 5 | 2608 | 72 | ||
NM_169769.2 | NP_732257.1 | 4 - 5 | 2516 | 72 | ||
NM_169770.3 | NP_732258.1 | 4 - 5 | 2578 | 72 | ||
NM_169771.2 | NP_732259.1 | 4 - 5 | 2523 | 72 | ||
NM_206505.2 | NP_996227.1 | 4 - 5 | 2521 | 72 | ||
GenBank mRNA | AY051768.1 | - | 5 - 6 | 2709 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |