Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144602.2 | NP_001138074.1 | 5 - 6 | 1105 | 70 | |
NM_001275775.1 | NP_001262704.1 | 5 - 6 | 1202 | 70 | ||
NM_001275776.1 | NP_001262705.1 | 5 - 6 | 1128 | 70 | ||
NM_142453.4 | NP_650710.3 | 5 - 6 | 1238 | 70 | ||
NM_169803.4 | NP_732325.3 | 5 - 6 | 1128 | 70 | ||
NM_169804.4 | NP_732326.3 | 5 - 6 | 1137 | 70 | ||
NM_169805.4 | NP_732327.3 | 5 - 6 | 1156 | 70 | ||
GenBank mRNA | AY118312.1 | - | 5 - 6 | 1238 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |