Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cona | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142462.3 | NP_650719.2 | 1 - 2 | 203 | 70 | |
GenBank mRNA | BT022998.1 | - | 1 - 2 | 203 | 70 | |
BT023086.1 | - | 1 - 2 | 203 | 70 | ||
BT023130.1 | - | 1 - 2 | 155 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |