Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mekk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142493.3 | NP_650750.4 | 5 - 6 | 5013 | 83 | |
NM_169833.2 | NP_732373.1 | 5 - 6 | 4807 | 83 | ||
GenBank mRNA | AY118659.1 | - | 5 - 6 | 4773 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |