Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mekk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142493.3 | NP_650750.4 | 6 - 7 | 5188 | 84 | |
NM_169833.2 | NP_732373.1 | 6 - 7 | 4982 | 84 | ||
GenBank mRNA | AY118659.1 | - | 6 - 7 | 4948 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |