Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp12a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142526.4 | NP_650783.2 | 1 - 2 | 123 | 100 | |
GenBank mRNA | BT056257.1 | - | 1 - 2 | 121 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |