Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpr92A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142556.3 | NP_650813.2 | 2 | 388 | 73 | |
GenBank mRNA | BT023042.1 | - | 2 | 382 | 73 | |
BT023126.1 | - | 2 | 388 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |