Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260274.2 | NP_001247203.1 | 3 - 4 | 1584 | 69 | |
NM_001275826.1 | NP_001262755.1 | 2 - 3 | 2294 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |