Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104377.2 | NP_001097847.1 | 9 - 10 | 6769 | 58 | |
NM_001275842.1 | NP_001262771.1 | 9 - 10 | 6769 | 58 | ||
NM_001275843.1 | NP_001262772.1 | 10 - 11 | 7042 | 58 | ||
GenBank mRNA | DQ494533.1 | - | 9 - 10 | 6680 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |