Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104378.2 | NP_001097848.2 | 9 - 10 | 1921 | 85 | |
NM_001104379.2 | NP_001097849.1 | 9 - 10 | 1844 | 85 | ||
NM_001275847.1 | NP_001262776.1 | 10 - 11 | 1931 | 85 | ||
NM_001275850.1 | NP_001262779.1 | 10 - 11 | 1931 | 85 | ||
NM_001275851.1 | NP_001262780.1 | 9 - 10 | 3256 | 85 | ||
NM_001275852.1 | NP_001262781.1 | 9 - 10 | 1670 | 85 | ||
NM_001275853.1 | NP_001262782.1 | 9 - 10 | 1921 | 85 | ||
NM_001275855.1 | NP_001262784.1 | 9 - 10 | 3256 | 85 | ||
NM_001369986.1 | NP_001356920.1 | 10 - 11 | 1931 | 85 | ||
NM_001369987.1 | NP_001356921.1 | 10 - 11 | 1931 | 85 | ||
NM_001369988.1 | NP_001356922.1 | 9 - 10 | 1670 | 85 | ||
NM_142656.4 | NP_650913.2 | 9 - 10 | 3256 | 85 | ||
NM_169925.3 | NP_732555.1 | 9 - 10 | 1670 | 85 | ||
NM_169926.2 | NP_732556.1 | 9 - 10 | 1670 | 85 | ||
NM_169927.3 | NP_732557.2 | 10 - 11 | 1931 | 85 | ||
NM_169928.3 | NP_732558.2 | 9 - 10 | 2370 | 85 | ||
NM_169929.3 | NP_732559.1 | 9 - 10 | 1921 | 85 | ||
GenBank mRNA | AY058477.1 | - | 9 - 10 | 2219 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |