Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nelf-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275875.1 | NP_001262804.1 | 3 - 4 | 2043 | 80 | |
NM_142701.2 | NP_650958.2 | 3 - 4 | 2043 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |