Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Burs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142726.2 | NP_650983.1 | 2 - 3 | 289 | 84 | |
GenBank mRNA | AJ862523.1 | - | 2 - 3 | 256 | 84 | |
AY672905.1 | - | 2 - 3 | 256 | 84 | ||
BT122042.1 | - | 2 - 3 | 256 | 84 | ||
BT132842.1 | - | 2 - 3 | 256 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |