Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp6d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142825.4 | NP_651082.1 | 1 - 2 | 767 | 135 | |
GenBank mRNA | AY051606.1 | - | 1 - 2 | 717 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |