Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275954.1 | NP_001262883.1 | 2 - 3 | 378 | 65 | |
NM_142926.4 | NP_651183.3 | 2 - 3 | 759 | 65 | ||
NM_170092.3 | NP_732904.2 | 2 - 3 | 378 | 65 | ||
NM_170093.3 | NP_732905.2 | 1 - 2 | 447 | 65 | ||
GenBank mRNA | AF357214.1 | - | 1 - 2 | 303 | 65 | |
AY051436.1 | - | 2 - 3 | 351 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |