Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142947.4 | NP_651204.1 | 1 - 2 | 178 | 76 | |
GenBank mRNA | AY113286.1 | - | 1 - 2 | 143 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |