Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316509.1 | NP_001303438.1 | 2 - 3 | 1220 | 60 | |
NM_142949.4 | NP_651206.3 | 2 - 3 | 1220 | 60 | ||
NM_170115.3 | NP_732941.2 | 2 - 3 | 1220 | 60 | ||
GenBank mRNA | AY069522.1 | - | 2 - 3 | 702 | 60 | |
BT044258.1 | - | 2 - 3 | 1217 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |